Gallussyre monohydrat
Leverandør: Thermo Scientific
Warning
Synonymer: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate
220761000 220765000 220760025
ACRO220761000EA 1000 NOK
ACRO220761000 ACRO220765000 ACRO220760025
Gallussyre monohydrat
Gallic acid monohydrate
Formel: C₇H₆O₅·1H₂O
MW: 188,14 g/mol
Smeltepunkt: 251 °C (dec.)
Flash punkt: 250 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149098
CAS-nummer: 5995-86-8
EINECS: 205-749-9
Bestill nå
Spesifikasjon Testresultater
Appearance Off-white to beige Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (120°C)
Sulfated ash ≤0.1 %
Titration with NaOH ≥99 % (On dry substance)