Adonitol ≥99%
Leverandør: Thermo Scientific
Synonymer:
(2R,3s,4S)-Pentane-1,2,3,4,5-pentol, Pentane-1,2,3,4,5-pentol, (2R,3s,4S)-Pentane-1,2,3,4,5-pentaol, Adonit, Ribitol, 1,2,3,4,5-Pentanol, 1,2,3,4,5-Pentanepentol, Adonite, Pentitol, Adonitrol
L03253.06
L03253.14
L03253.06EA
550
NOK
L03253.06
L03253.14
Adonitol ≥99%
Adonitol
Purity: 99%
CAS No.: 488-81-3
Molecular Formula: C5H12O5
Formula Weight: 152.15
Melting Point: 102-104°
Beilstein Reference: 1720524
Merck Reference: 14,168
MDL No.: MFCD00064291
Formel:
HOH₂C(CH(OH))₃CH₂OH MW: 152,15 g/mol Smeltepunkt: 102…104 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00064291 CAS-nummer: 488-81-3 EINECS: 207-685-7 Merck Indeks: 14,168 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...