4-Nitrobenzyl acetate ≥98.0% (by total nitrogen basis)

Leverandør: TCI

Synonymer: Acetic Acid 4-Nitrobenzyl Ester

A0039-25G A0039-5G
TCIAA0039-25GEA 1375 NOK
TCIAA0039-25G TCIAA0039-5G
4-Nitrobenzyl acetate ≥98.0% (by total nitrogen basis)
4-Nitrobenzyl acetate
Formel: C₉H₉NO₄
MW: 195,17 g/mol
MDL nummer: MFCD00024781
CAS-nummer: 619-90-9
EINECS: 210-618-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 76.0 to 79.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR