Aluminon mixture of isomers

Leverandør: TCI
Warning

Synonymer: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt

A0240-25G A0240-5G
TCIAA0240-25GEA 3400 NOK
TCIAA0240-25G TCIAA0240-5G
Aluminon mixture of isomers
Aluminon
Formel: C₂₂H₂₃N₃O₉
MW: 473,44 g/mol
MDL nummer: MFCD00040925
CAS-nummer: 569-58-4
EINECS: 209-319-1
Merck Indeks: 14,00322

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Sensitiveness Colorimetric Test : Turn Red with 1ppm Aluminium

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR