Disodium adipate ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
Disodium Adipate, Sodium adipate, Adipic Acid Disodium Salt
A0592-25G
A0592-500G
TCIAA0592-25GEA
555
NOK
TCIAA0592-25G
TCIAA0592-500G
Disodium adipate ≥98.0% (by GC, titration analysis)
Disodium adipate
Formel:
C₆H₁₀O₄ MW: 190,11 g/mol |
MDL nummer:
MFCD00070498 CAS-nummer: 7486-38-6 EINECS: 231-293-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...