4-Aminocinnamic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 3-(4-Aminophenyl)acrylic acid, 4-Aminocinnamic Acid, 3-(4-Aminophenyl)acrylic Acid

A0691-25G A0691-5G
TCIAA0691-25GEA 4100 NOK
TCIAA0691-25G TCIAA0691-5G
4-Aminocinnamic acid ≥98.0% (by HPLC, titration analysis)
4-Aminocinnamic acid
Formel: C₉H₉NO₂
MW: 163,18 g/mol
MDL nummer: MFCD00017117
CAS-nummer: 2393-18-2
EINECS: 219-246-7

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in dilute HCl almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR