Sodium-4-aminoazobenzene-4'-sulphonate ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
4-Aminoazobenzene-4'-sulphonic acid Sodium Salt
A0748-500G
A0748-25G
TCIAA0748-500GEA
2425
NOK
TCIAA0748-500G
TCIAA0748-25G
Sodium-4-aminoazobenzene-4'-sulphonate ≥98.0% (by HPLC)
Sodium-4-aminoazobenzene-4'-sulphonate
Formel:
C₁₂H₁₁N₃O₃S MW: 299,28 g/mol |
MDL nummer:
MFCD00035564 CAS-nummer: 2491-71-6 EINECS: 219-657-1 |
Spesifikasjon Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...