Acetone dibutyl acetal ≥95.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2,2-Dibutoxypropane
A0782-25ML
A0782-5ML
TCIAA0782-25MLEA
1875
NOK
TCIAA0782-25ML
TCIAA0782-5ML
Acetone dibutyl acetal ≥95.0% (by GC)
Acetone dibutyl acetal
Formel:
C₁₁H₂₄O₂ MW: 188,31 g/mol Kokepunkt: 130°C |
MDL nummer:
MFCD00015248 CAS-nummer: 141-72-0 EINECS: 205-496-4 FN: 3271 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...