Isoamyl acetoacetate ≥97.0% (by GC)

Leverandør: TCI
Warning

A0812-25ML A0812-500ML
TCIAA0812-25MLEA 540 NOK
TCIAA0812-25ML TCIAA0812-500ML
Isoamyl acetoacetate ≥97.0% (by GC)
Isoamyl acetoacetate
Formel: C₉H₁₆O₃
MW: 172,22 g/mol
Kokepunkt: 218°C
Flash punkt: 101°C
MDL nummer: MFCD00059355
CAS-nummer: 2308-18-1
EINECS: 218-991-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR