4-Acryloylmorpholine ≥98.0% (by GC) stabilised

Leverandør: TCI
Danger


Stabilisering: Stabilised with 4-methoxyphenol (MEHQ)

A0841-25ML A0841-500ML A0841-100ML
TCIAA0841-25MLEA 230 NOK
TCIAA0841-25ML TCIAA0841-500ML TCIAA0841-100ML
4-Acryloylmorpholine ≥98.0% (by GC) stabilised
4-Acryloylmorpholine
Formel: C₇H₁₁NO₂
MW: 141,17 g/mol
Kokepunkt: 165°C
Flash punkt: 127°C
MDL nummer: MFCD00047413
CAS-nummer: 5117-12-4
EINECS: 418-140-1

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR