Cinnamyl acetate ≥97.0% (by GC)

Leverandør: TCI

Synonymer: Acetic Acid Cinnamyl Ester

A0901-25ML A0901-500ML
TCIAA0901-25MLEA 220 NOK
TCIAA0901-25ML TCIAA0901-500ML
Cinnamyl acetate ≥97.0% (by GC)
Cinnamyl acetate
Formel: C₁₁H₁₂O₂
MW: 176,22 g/mol
Flash punkt: 118°C
MDL nummer: MFCD00008722
CAS-nummer: 103-54-8
EINECS: 203-121-9

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR