Angelic anhydride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Angelic Anhydride, ['(', 'Z', ')-2-Methyl-2-butenoic Anhydride', '2-Methylisocrotonic Anhydride'], (Z)-2-Methyl-2-butenoic Anhydride, 2-Methylisocrotonic Anhydride

A1180-1G A1180-5G
TCIAA1180-1GEA 3025 NOK
TCIAA1180-1G TCIAA1180-5G
Angelic anhydride ≥98.0% (by titrimetric analysis)
Angelic anhydride
Formel: C₁₀H₁₄O₃
MW: 182,22 g/mol
Kokepunkt: 108°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00198080
CAS-nummer: 94487-74-8
FN: 3265
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Morpholine Method) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR