Allylsuccinic anhydride ≥97.0% (by GC)

Leverandør: TCI
Danger

Synonymer: (2-Propenyl)butanedioic Anhydride, 4-Pentene-1,2-dicarboxylic Anhydride

A1283-25G A1283-5G
TCIAA1283-25GEA 1825 NOK
TCIAA1283-25G TCIAA1283-5G
Allylsuccinic anhydride ≥97.0% (by GC)
Allylsuccinic anhydride
Formel: C₇H₈O₃
MW: 140,14 g/mol
Kokepunkt: 143°C
MDL nummer: MFCD00085227
CAS-nummer: 7539-12-0
EINECS: 231-412-0
FN: 2810
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR