(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
SAMP
A1335-1G
A1335-5G
TCIAA1335-1GEA
2250
NOK
TCIAA1335-1G
TCIAA1335-5G
(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine ≥98.0% (by titrimetric analysis)
(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine
Formel:
C₆H₁₄N₂O MW: 130,19 g/mol Kokepunkt: 64°C Flash punkt: 72°C |
MDL nummer:
MFCD00064485 CAS-nummer: 59983-39-0 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -78.0 to -84.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...