4-Aminononafluorobiphenyl ≥96.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Perfluoro-4-aminobiphenyl, 2,2',3,3',4',5,5',6,6'-Nonafluoro-[1,1'-biphenyl]-4-amine

A1346-5G A1346-1G
TCIAA1346-5GEA 4000 NOK
TCIAA1346-5G TCIAA1346-1G
4-Aminononafluorobiphenyl ≥96.0% (by GC)
4-Aminononafluorobiphenyl
Formel: C₁₂H₂F₉N
MW: 331,14 g/mol
MDL nummer: MFCD00671483
CAS-nummer: 969-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=660c06d6bddef704946d3530b47dd359e662f85b&vid=660c06d6bddef704946d3530b47dd359e662f85b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 144.0 to 148.0 deg-C
Solubility in Toluene within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR