Aminoacetamidine dihydrobromide ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Warning

A1361-1G A1361-5G
TCIAA1361-1GEA 1125 NOK
TCIAA1361-1G TCIAA1361-5G
Aminoacetamidine dihydrobromide ≥95.0% (by titrimetric analysis)
Aminoacetamidine dihydrobromide
Formel: C₂H₇N₃
MDL nummer: MFCD00671486
CAS-nummer: 69816-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5c6ab00ae3139c4f432ddc1e5f9e1c0d2024a85&vid=d5c6ab00ae3139c4f432ddc1e5f9e1c0d2024a85&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR