1-Methyl-2-aminoterephthalate ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Amino-4-methoxycarbonylbenzoic Acid, 3-Amino-4-carbomethoxybenzoic Acid, 2-Aminoterephthalic Acid 1-Methyl Ester
A1507-25G
A1507-5G
TCIAA1507-25GEA
1775
NOK
TCIAA1507-25G
TCIAA1507-5G
1-Methyl-2-aminoterephthalate ≥98.0% (by titrimetric analysis)
1-Methyl-2-aminoterephthalate
Formel:
C₉H₉NO₄ MW: 195.17 g/mol |
MDL nummer:
MFCD00189374 CAS-nummer: 60728-41-8 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 216.0 to 221.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...