(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
(1R,2S)-(+)-cis-1-Amino-2-indanol, (1R,2S)-(+)-cis-1-Aminoindan-2-ol
A1623-1G
A1623-5G
TCIAA1623-1GEA
220
NOK
TCIAA1623-1G
TCIAA1623-5G
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol
Formel:
C₉H₁₁NO MW: 149,19 g/mol |
MDL nummer:
MFCD00216656 CAS-nummer: 136030-00-7 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...