2,3-Anthracenedicarboxylic anhydride ≥93.0% (by GC)

Leverandør: TCI
Warning

A1834-100MG A1834-500MG
TCIAA1834-100MGEA 1350 NOK
TCIAA1834-100MG TCIAA1834-500MG
2,3-Anthracenedicarboxylic anhydride ≥93.0% (by GC)
2,3-Anthracenedicarboxylic anhydride
Formel: C₁₆H₈O₃
MW: 248,24 g/mol
CAS-nummer: 6812-14-2

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR