2-Acetamido-6-nitrobenzoic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Acetamido-6-nitrobenzoic Acid, 2'-Carboxy-3'-nitroacetanilide, N-Acetyl-6-nitroanthranilic acid, 2-(Acetylamino)-6-nitrobenzoic Acid, N-Acetyl-6-nitroanthranilic Acid

A1841-5G A1841-1G
TCIAA1841-5GEA 1675 NOK
TCIAA1841-5G TCIAA1841-1G
2-Acetamido-6-nitrobenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Acetamido-6-nitrobenzoic acid
Formel: C₉H₈N₂O₅
MW: 224,17 g/mol
MDL nummer: MFCD08276267
CAS-nummer: 73721-78-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR