3A-Amino-3A-deoxy-(2AS,3AS)-ɑ-cyclodextrin hydrate ≥90.0% (by HPLC)

Leverandør: TCI

Synonymer: 3A-Amino-3A-deoxy-(2AS,3AS)-α-cyclodextrin Hydrate

A2122-1G A2122-200MG
TCIAA2122-1GEA 2975 NOK
TCIAA2122-1G TCIAA2122-200MG
3A-Amino-3A-deoxy-(2AS,3AS)-ɑ-cyclodextrin hydrate ≥90.0% (by HPLC)
3A-Amino-3A-deoxy-(2AS,3AS)-alpha-cyclodextrin hydrate
Formel: C₃₆H₆₁NO₂₉
MW: 971,86 g/mol
CAS-nummer: 121916-94-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10d91c36c704665cc4a81ff04de7a8023e40cf4e&vid=10d91c36c704665cc4a81ff04de7a8023e40cf4e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 90.0 area%
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +127 to +138 deg (C=1, H2O) (calcd.on anh.substance)
Water max. 15.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR