(S)-2-Aminobutanamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
|
Warning
|
Synonymer:
H-Abu-NH2 · HCl, (2S)-2-Aminobutanamide hydrochloride, (S)-(+)-2-Aminobutanamide hydrochloride, H-Abu-NH2 HCl
A2252-25G
A2252-5G
TCIAA2252-25GEA
715
NOK
TCIAA2252-25G
TCIAA2252-5G
(S)-2-Aminobutanamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
(S)-2-Aminobutanamide hydrochloride
Formel:
C₄H₁₁ClN₂O MW: 138,60 g/mol |
MDL nummer:
MFCD00136565 CAS-nummer: 7682-20-4 EINECS: 471-490-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | +23.0 to +25.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...