2-Amino-5-methylthiophene-3-carbonitrile ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Amino-5-methyl-3-thiophenecarbonitrile, 2-Amino-3-cyano-5-methylthiophene
A2351-25G
A2351-5G
TCIAA2351-25GEA
2550
NOK
TCIAA2351-25G
TCIAA2351-5G
2-Amino-5-methylthiophene-3-carbonitrile ≥98.0% (by GC)
2-Amino-5-methylthiophene-3-carbonitrile
Formel:
C₆H₆N₂S MW: 138,1902 g/mol |
MDL nummer:
MFCD01626055 CAS-nummer: 138564-58-6 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...