2-Amino-4-methoxybenzothiazole ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 4-Methoxybenzothiazol-2-ylamine

A2454-1G A2454-5G
TCIAA2454-1GEA 755 NOK
TCIAA2454-1G TCIAA2454-5G
2-Amino-4-methoxybenzothiazole ≥98.0% (by HPLC)
2-Amino-4-methoxybenzothiazole
Formel: C₈H₈N₂OS
MW: 180,23 g/mol
MDL nummer: MFCD00005792
CAS-nummer: 5464-79-9
EINECS: 226-763-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR