(-)-Ambroxide ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Ambroxide, (3aR,5aS,9aS,9bR)-3a,6,6,9a-Tetramethyldodecahydronaphtho[2,1-b]furan, 1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane
A2472-5G
A2472-25G
TCIAA2472-5GEA
865
NOK
TCIAA2472-5G
TCIAA2472-25G
(-)-Ambroxide ≥98.0% (by GC)
(-)-Ambroxide
Formel:
C₁₆H₂₈O MW: 236,40 g/mol |
MDL nummer:
MFCD00134491 CAS-nummer: 6790-58-5 EINECS: 229-861-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Specific rotation [a]20/D | -24.0 to -27.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...