Aripiprazole ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone

A2496-1G A2496-5G
TCIAA2496-1GEA 1025 NOK
TCIAA2496-1G TCIAA2496-5G
Aripiprazole ≥98.0% (by HPLC, titration analysis)
Aripiprazole
Formel: C₂₃H₂₇Cl₂N₃O₂
MW: 448,39 g/mol
MDL nummer: MFCD00892072
CAS-nummer: 129722-12-9
Merck Indeks: 14,00785

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR