1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid ≥95.0% (by HPLC)

Leverandør: TCI

Synonymer: Amino-PEG6-carboxylate, 21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic acid, Amino-PEG6-carboxylic acid, NH2-PEG6-CH2CH2COOH, NH2-PEG6-COOH, 1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid, Carboxy-PEG6-amine, 21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic Acid

A2526-100MG A2526-500MG
TCIAA2526-100MGEA 1525 NOK
TCIAA2526-100MG TCIAA2526-500MG
1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid ≥95.0% (by HPLC)
1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid
Formel: C₁₅H₃₁NO₈
MW: 353,41 g/mol
MDL nummer: MFCD13184943
CAS-nummer: 905954-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2cedb8779246e4d2761615a762ff5abc89e1524&vid=ea3ea52856ac4cc8e0bfe3e12b5d6affa99510c5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 95.0 area%
Elemental analysis (Nitrogen) 3.60 to 4.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR