2-(Aminomethyl)-1-ethylpyrrolidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

A2556-5G A2556-25G
TCIAA2556-5GEA 825 NOK
TCIAA2556-5G TCIAA2556-25G
2-(Aminomethyl)-1-ethylpyrrolidine ≥98.0% (by GC, titration analysis)
2-(Aminomethyl)-1-ethylpyrrolidine
Formel: C₇H₁₆N₂
MW: 128,22 g/mol
Kokepunkt: 60°C
Flash punkt: 56°C
MDL nummer: MFCD00003178
CAS-nummer: 26116-12-1
EINECS: 247-466-3
FN: 2733
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR