2-Amino-5-bromo-3-iodopyridine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
5-Bromo-3-iodo-2-pyridylamine, 5-Bromo-3-iodopyridin-2-amine
A2563-1G
A2563-5G
TCIAA2563-1GEA
690
NOK
TCIAA2563-1G
TCIAA2563-5G
2-Amino-5-bromo-3-iodopyridine ≥98.0% (by titrimetric analysis)
2-Amino-5-bromo-3-iodopyridine
Formel:
C₅H₄BrIN₂ MW: 298,91 g/mol |
MDL nummer:
MFCD06659000 CAS-nummer: 381233-96-1 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | gt. 112.0 and le 116.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...