ɑ-Asarone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: alpha-Asarone, trans-1,2,4-Trimethoxy-5-(1-propenyl)benzene, (E)-1,2,4-Trimethoxy-5-(prop-1-en-1-yl)benzene

A2590-1G A2590-5G
TCIAA2590-1GEA 405 NOK
TCIAA2590-1G TCIAA2590-5G
ɑ-Asarone ≥98.0% (by GC)
alpha-Asarone
Formel: C₁₂H₁₆O₃
MW: 208,26 g/mol
Kokepunkt: 298°C
MDL nummer: MFCD00064457
CAS-nummer: 2883-98-9
EINECS: 220-743-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR