4-(trans-4-Amylcyclohexyl)benzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-(trans-4-Pentylcyclohexyl)benzoic Acid
A2593-5G
A2593-25G
TCIAA2593-5GEA
485
NOK
TCIAA2593-5G
TCIAA2593-25G
4-(trans-4-Amylcyclohexyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-(trans-4-Amylcyclohexyl)benzoic acid
Formel:
C₁₈H₂₆O₂ MW: 274,40 g/mol |
MDL nummer:
MFCD06208360 CAS-nummer: 65355-30-8 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 179.0 and le 183.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...