5-Aminobenzimidazole ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Amino-1H-benzimidazole

A2605-1G A2605-5G
TCIAA2605-1GEA 660 NOK
TCIAA2605-1G TCIAA2605-5G
5-Aminobenzimidazole ≥98.0% (by GC)
5-Aminobenzimidazole
Formel: C₇H₇N₃
MW: 133,15 g/mol
Kokepunkt: 222°C
MDL nummer: MFCD00465258
CAS-nummer: 934-22-5
EINECS: 213-279-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18acb7f4b92991326089fed3c46d710062b1db48&vid=324b819dda7750bf72a5a77f36292a2abd9786f6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 162.0 to 167.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR