Anisomycin ≥97.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
(2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate
A2613-25MG
A2613-100MG
TCIAA2613-25MGEA
1675
NOK
TCIAA2613-25MG
TCIAA2613-100MG
Anisomycin ≥97.0% (by GC)
Anisomycin
Formel:
C₁₄H₁₉NO₄ MW: 265,31 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD00077650 CAS-nummer: 22862-76-6 EINECS: 245-269-7 FN: 3462 ADR: 6.1,III Merck Indeks: 14,00670 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 141.0 to 145.0 deg-C |
Specific rotation [a]20/D | -29.0 to -31.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...