4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid

A2615-1G A2615-5G
TCIAA2615-1GEA 395 NOK
TCIAA2615-1G TCIAA2615-5G
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic acid
Formel: C₁₀H₁₃NO₅S
MW: 259,28 g/mol
MDL nummer: MFCD04973619
CAS-nummer: 71675-87-1
EINECS: 275-833-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 148.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR