2-Amino-5-(benzylthio)-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
5-(Benzylthio)-1,3,4-thiadiazol-2-amine, 5-(Benzylthio)-1,3,4-thiadiazole-2-amine
A2677-1G
A2677-5G
TCIAA2677-1GEA
1375
NOK
TCIAA2677-1G
TCIAA2677-5G
2-Amino-5-(benzylthio)-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-(benzylthio)-1,3,4-thiadiazole
Formel:
C₉H₉N₃S₂ MW: 223,31 g/mol |
MDL nummer:
MFCD00184237 CAS-nummer: 25660-71-3 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystalline |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...