2-Allylisoindoline-1,3-dione ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
N-Allylphthalimide
A2696-200MG
A2696-1G
TCIAA2696-200MGEA
555
NOK
TCIAA2696-200MG
TCIAA2696-1G
2-Allylisoindoline-1,3-dione ≥98.0% (by GC)
2-Allylisoindoline-1,3-dione
Formel:
C₁₁H₉NO₂ MW: 187,19 g/mol Kokepunkt: 295°C |
MDL nummer:
MFCD00158662 CAS-nummer: 5428-09-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...