AG-494 ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
|
Danger
|
A2704-20MG
A2704-100MG
TCIAA2704-20MGEA
1000
NOK
TCIAA2704-20MG
TCIAA2704-100MG
AG-494 ≥98.0% (by HPLC, total nitrogen)
AG-494
Formel:
C₁₆H₁₂N₂O₃ Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00209863 CAS-nummer: 133550-35-3 FN: 3439 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...