4-Amino-2-chloropyrimidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Chlor-4-pyrimidinamin, 2-Chloro-4-pyrimidinamine, 2-Chloropyrimidin-4-amine, 2-Chloropyrimidin-4-ylamine

A2719-1G A2719-5G
TCIAA2719-1GEA 445 NOK
TCIAA2719-1G TCIAA2719-5G
4-Amino-2-chloropyrimidine ≥98.0% (by GC, titration analysis)
4-Amino-2-chloropyrimidine
Formel: C₄H₄ClN₃
MW: 129,55 g/mol
MDL nummer: MFCD00194051
CAS-nummer: 7461-50-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR