4-Amino-2-methoxybenzonitrile ≥97.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
A2785-1G
A2785-5G
TCIAA2785-1GEA
1125
NOK
TCIAA2785-1G
TCIAA2785-5G
4-Amino-2-methoxybenzonitrile ≥97.0% (by GC, titration analysis)
4-Amino-2-methoxybenzonitrile
Formel:
C₈H₈N₂O MW: 148,1619 g/mol |
MDL nummer:
MFCD09027303 CAS-nummer: 7251-09-4 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...