4-Amino-3-chloropyridine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

A2788-1G A2788-5G
TCIAA2788-1GEA 555 NOK
TCIAA2788-1G TCIAA2788-5G
4-Amino-3-chloropyridine ≥97.0% (by GC, titration analysis)
4-Amino-3-chloropyridine
Formel: C₅H₅ClN₂
MW: 128,56 g/mol
MDL nummer: MFCD03984473
CAS-nummer: 19798-77-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR