1-(Azidomethyl)pyrene ≥98.0% (by HPLC)

Leverandør: TCI

A2791-200MG A2791-1G
TCIAA2791-200MGEA 1500 NOK
TCIAA2791-200MG TCIAA2791-1G
1-(Azidomethyl)pyrene ≥98.0% (by HPLC)
1-(Azidomethyl)pyrene
Formel: C₁₇H₁₁N₃
MW: 257,30 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 1006061-57-9

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 15.50 to 17.00 %
Melting point 66.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR