7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: AMCA, 2H-1-Benzopyran-3-acetic acid, 7-Amino-4-methylcoumarin-3-acetic acid, Aminomethyl coumarin acetic acid, 2-(7-Amino-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid

A2820-50MG A2820-250MG
TCIAA2820-50MGEA 485 NOK
TCIAA2820-50MG TCIAA2820-250MG
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95.0% (by titrimetric analysis)
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H)
Formel: C₁₂H₁₁NO₄
MW: 233,22 g/mol
MDL nummer: MFCD00081051
CAS-nummer: 106562-32-7

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 93.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR