4-Amino-o-anisic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 4-Amino-2-methoxybenzenecarboxylic acid

A2840-5G A2840-25G
TCIAA2840-5GEA 555 NOK
TCIAA2840-5G TCIAA2840-25G
4-Amino-o-anisic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-o-anisic acid
Formel: C₈H₉NO₃
MW: 167,16 g/mol
MDL nummer: MFCD00114479
CAS-nummer: 2486-80-8
EINECS: 219-632-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR