4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Amino-1-methyl-3-N-propyl-1H-pyrazole-5-carboxamide, 4-Amino-1-methyl-3-propylpyrazole-5-carboxamide
A2857-5G
A2857-25G
TCIAA2857-5GEA
500
NOK
TCIAA2857-5G
TCIAA2857-25G
4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide ≥98.0% (by HPLC, titration analysis)
4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide
Formel:
C₈H₁₄N₄O MW: 182,23 g/mol |
MDL nummer:
MFCD02927682 CAS-nummer: 139756-02-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...