9-Aminoacridine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 9-AA

A2905-1G A2905-5G
TCIAA2905-1GEA 540 NOK
TCIAA2905-1G TCIAA2905-5G
9-Aminoacridine ≥97.0% (by GC, titration analysis)
9-Aminoacridine
Formel: C₁₃H₁₀N₂
MW: 194,24 g/mol
MDL nummer: MFCD00037839
CAS-nummer: 90-45-9
EINECS: 201-995-6
Merck Indeks: 14,00407

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 236.0 to 240.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR