2-Amino-5-chloro-6-methylpyridine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 5-Chloro-6-methylpyridin-2-amine

A2920-1G A2920-5G
TCIAA2920-1GEA 755 NOK
TCIAA2920-1G TCIAA2920-5G
2-Amino-5-chloro-6-methylpyridine ≥98.0% (by titrimetric analysis)
2-Amino-5-chloro-6-methylpyridine
Formel: C₆H₇ClN₂
MW: 142,59 g/mol
MDL nummer: MFCD09453335
CAS-nummer: 36936-23-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR