Azido-PEG4-Amine ≥95.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Amino-PEG4-Azide, 14-Amino-3,6,9,12-tetraoxatetradecanyl Azide, 14-Azido-3,6,9,12-tetraoxatetradecan-1-amine, 1-Amino-14-azido-3,6,9,12-tetraoxatetradecane

A3004-50MG A3004-250MG
TCIAA3004-50MGEA 610 NOK
TCIAA3004-50MG TCIAA3004-250MG
Azido-PEG4-Amine ≥95.0% (by GC)
Azido-PEG4-Amine
Formel: C₁₀H₂₂N₄O₄
MW: 262,31 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD20134134
CAS-nummer: 951671-92-4
FN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32fca75e2e755551477537904f299fb62a7f8ac&vid=e32fca75e2e755551477537904f299fb62a7f8ac&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR