3-Oxetanamine ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-amino-oxetane, 3-Aminooxetane

A3080-200MG A3080-1G
TCIAA3080-200MGEA 540 NOK
TCIAA3080-200MG TCIAA3080-1G
3-Oxetanamine ≥97.0% (by GC)
3-Oxetanamine
Formel: C₃H₇NO
MW: 73,09 g/mol
Flash punkt: 54°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD08544363
CAS-nummer: 21635-88-1
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74625f81eb93be0f751ce84cdfadf7e961c6bfd2&vid=74625f81eb93be0f751ce84cdfadf7e961c6bfd2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR