AM 580 ≥97.0% (by GC)

Leverandør: TCI

Synonymer: NSC 608001, CD336, 4-​​[[(5,​​6,​​7,​​8-​​tetrahydro-​​5,​​5,​​8,​​8-​​tetramethyl-​​2-​​naphthalenyl)​carbonyl]​amino]​-​benzoic acid, AM580, 4-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxamido)benzoic acid, RO 40-6055

A3082-100MG A3082-25MG A3082-5MG
TCIAA3082-100MGEA 7075 NOK
TCIAA3082-100MG TCIAA3082-25MG TCIAA3082-5MG
AM 580 ≥97.0% (by GC)
AM 580
Formel: C₂₂H₂₅NO₃
MW: 351.44 g/mol
MDL nummer: MFCD00673916
CAS-nummer: 102121-60-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR