Amodiaquine dihydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: Amodiaquin dihydrochloride dihydrate, 4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate, 4-([7-Chloro-4-quinolinyl]amino)-2-([diethylamino]methyl)phenol, 4-[(7-Chloroquinolin-4-yl)amino]-2-[(diethylamino)methyl]phenol dihydrochloride

A3133-5G A3133-25G
TCIAA3133-5GEA 245 NOK
TCIAA3133-5G TCIAA3133-25G
Amodiaquine dihydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
Amodiaquine dihydrochloride dihydrate
Formel: C₂₀H₂₂ClN₃O·2HCl·2H₂O
MW: 464,812 g/mol (392,33 anhy)
MDL nummer: MFCD00078857
CAS-nummer: 6398-98-7
EINECS: 200-706-0
Merck Indeks: 14,00572
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea34b7af3f0b52d5a0c504e3dd62f70a51182ebf&vid=ea34b7af3f0b52d5a0c504e3dd62f70a51182ebf&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 7.0 to 9.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR